Learn More
Bioworld 2'-Deoxycytidine Hydrochloride, 10 g
Supplier:  Bioworld 404900042
Encompass_Preferred
Specifications:Ultra Pure GradeEm (280 nm, 0.1N HCl): >12,800Hygroscopic Appearance: White crystalline powderChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 168-170°C
Catalog No. 50-255-895