Learn More
Medchemexpress LLC 2'-Deoxycytidine hydrochloride | 3992-42-5 | 100.0% | 263.68 | 500 MG
SDP
Catalog No. 5000387217
Encompass_Preferred
2'-Deoxycytidine hydrochloride is an orally effective inhibitor of nucleic acid biosynthesis enzymes. It works by competing with these enzymes and nucleoside transporters, which helps to reduce the toxicity associated with cytarabine. This compound is also utilized in research related to L1210 lymphoblastic leukemia.
  • Orally effective nucleic acid biosynthesis enzyme inhibitor.
  • Competes with nucleoside transporters.
  • Reduces cytarabine toxicity.
  • Applicable in L1210 lymphoblastic leukemia research.
Catalog No. 50-003-87217 Supplier Medchemexpress LLC Supplier No. HY17564500MG
2'-Deoxycytidine hydrochloride is an orally effective inhibitor of nucleic acid biosynthesis enzymes. It works by competing with these enzymes and nucleoside transporters, which helps to reduce the toxicity associated with cytarabine. This compound is also utilized in research related to L1210 lymphoblastic leukemia.
  • Orally effective nucleic acid biosynthesis enzyme inhibitor.
  • Competes with nucleoside transporters.
  • Reduces cytarabine toxicity.
  • Applicable in L1210 lymphoblastic leukemia research.