Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Deoxycytidine hydrochloride, 99%, MilliporeSigma™ Supelco™

Catalog No. 111014138 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4138 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4138 Supplier MilliporeSigma Supelco Supplier No. D800610G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 3992-42-5
Physical Form Neat
Linear Formula C9H13N3O4 · HCl
MDL Number MFCD00012840
Synonym 1-(2-Deoxy-beta-D-ribofuranosyl)cytosine hydrochloride; Cytosine deoxyriboside hydrochloride
Type Analytical Standard
Quantity 10 g
Formula Weight 263.68
Percent Purity ≥99%
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.