Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Deoxycytidine Hydrochloride, Synthetic, Thermo Scientific™

Catalog No. AAJ1428409
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1428409 10 g Plastic bottle
AAJ1428422 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ1428409 Supplier Thermo Scientific Supplier No. J1428409

Chemical Identifiers

CAS 3992-42-5
Molecular Formula C9H14ClN3O4
Molecular Weight (g/mol) 263.68
MDL Number MFCD00012840
InChI Key LTKCXZGFJFAPLY-VOBHXVAMNA-N
Synonym 2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl
PubChem CID 107488
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Melting Point ∼170°C (decomposition)
Packaging Plastic bottle
Quantity 10 g
Beilstein 3576124
Formula Weight 263.68
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxycytidine hydrochloride
EINECSNumber 223-639-9
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.