Learn More
Medchemexpress LLC 2'-Deoxycytidine (Standard) | 951-77-9 | 100.0% | 100 MG
SDP
Catalog No. 5000409802
Encompass_Preferred
2'-Deoxycytidine (Standard) is an endogenous metabolite and reference standard. It has a molecular weight of 227.22 and a formula of C9H13N3O4. It appears as a white to off-white solid and is intended for research and analytical applications.
  • Used as an analytical standard and reference standard for assays.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Applications include HPLC, GC, and MS.
  • Also classified as an endogenous metabolite and natural product.
Catalog No. 50-004-09802 Supplier Medchemexpress LLC Supplier No. HYD0184R100MG
2'-Deoxycytidine (Standard) is an endogenous metabolite and reference standard. It has a molecular weight of 227.22 and a formula of C9H13N3O4. It appears as a white to off-white solid and is intended for research and analytical applications.
  • Used as an analytical standard and reference standard for assays.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Applications include HPLC, GC, and MS.
  • Also classified as an endogenous metabolite and natural product.