Learn More
Cayman Chemical 2'-Deoxycytidine (hydrochloride), Purity: ≥98%, CAS: 3992-42-5, 1 g
SDP
Catalog No. 502988617
Encompass_Preferred
A deoxyribonucleoside; phosphorylated by deoxycytidine kinase in the cytosol or TK2 in mitochondria to form dCMP; oral administration in combination with deoxythymidine delays disease onset and increases lifespan in a mouse model of TK2 deficiency
Catalog No. 50-298-8617 Supplier Cayman Chemical Supplier No. 301251 G
A deoxyribonucleoside; phosphorylated by deoxycytidine kinase in the cytosol or TK2 in mitochondria to form dCMP; oral administration in combination with deoxythymidine delays disease onset and increases lifespan in a mouse model of TK2 deficiency