Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyguanosine-13C monohydrate | 478511-26-1 | MFCD00080300 | 99.9% | 286.25 g/mol | C9(13)CH15N5O5 | 10 MG
SDP

Catalog No. 5000281846
Encompass_Preferred

2'-Deoxyguanosine-13C monohydrate is the carbon-13 isotopically labeled form of 2'-deoxyguanosine provided as the monohydrate solid for analytical and research applications, including metabolic flux analysis and isotope-tracing studies. The material is supplied with documentation (COA, SDS) and is intended for research use only.

  • 13C labeled 2'-deoxyguanosine.
  • Monohydrate solid form.
  • Purity: 99.9% (HPLC).
  • Molecular weight: 286.25 g/mol.
  • Chemical formula: C9(13)CH15N5O5.
  • CAS number: 478511-26-1.
  • Typical pack size: 10 mg.
  • Documentation available: COA and SDS for handling and storage.
  • For research use only; not for human use.

Catalog No. 50-002-81846 Supplier Medchemexpress LLC Supplier No. HYW338294S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyguanosine-13C monohydrate is the carbon-13 isotopically labeled form of 2'-deoxyguanosine provided as the monohydrate solid for analytical and research applications, including metabolic flux analysis and isotope-tracing studies. The material is supplied with documentation (COA, SDS) and is intended for research use only.

  • 13C labeled 2'-deoxyguanosine.
  • Monohydrate solid form.
  • Purity: 99.9% (HPLC).
  • Molecular weight: 286.25 g/mol.
  • Chemical formula: C9(13)CH15N5O5.
  • CAS number: 478511-26-1.
  • Typical pack size: 10 mg.
  • Documentation available: COA and SDS for handling and storage.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.