Learn More
Medchemexpress LLC 2'-Deoxyguanosine-13C10,15N5 | 872624-05-0 | 282.13 | 1 MG
SDP
Catalog No. 5000387111
Encompass_Preferred
2'-Deoxyguanosine-13C10,15N5 is a 13C- and 15N-labeled 2'-Deoxyguanosine for research use only. This compound is used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process, with deuteration showing potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-87111 Supplier Medchemexpress LLC Supplier No. HY17563S11MG
2'-Deoxyguanosine-13C10,15N5 is a 13C- and 15N-labeled 2'-Deoxyguanosine for research use only. This compound is used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process, with deuteration showing potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS