Learn More
Medchemexpress LLC 2'-Deoxyguanosine-13C10,15N5 monohydrate | 2483830-26-6 | 99.8% | 1 MG
SDP
Catalog No. 5000410947
Encompass_Preferred
2'-Deoxyguanosine-13C10,15N5 (monohydrate) is the 13C and 15N labeled 2'-Deoxyguanosine monohydrate, which is an endogenous metabolite. It is for research use only and not sold to patients.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-10947 Supplier Medchemexpress LLC Supplier No. HYD0185S41MG
2'-Deoxyguanosine-13C10,15N5 (monohydrate) is the 13C and 15N labeled 2'-Deoxyguanosine monohydrate, which is an endogenous metabolite. It is for research use only and not sold to patients.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.