Learn More
Medchemexpress LLC 2′-deoxyguanosine 5′-monophosphate-13C10,15N5 dilithium | MFCD00057076 | 99.8% | 373.98 g/mol | 13C10H12Li215N5O7P | 10 MG
SDP
Catalog No. 5000281487
Encompass_Preferred
2′-Deoxyguanosine 5′-monophosphate-13C10,15N5 dilithium is an isotopically labeled nucleotide provided as the dilithium salt. It is intended for use as a tracer and internal standard in quantitative analyses by NMR, GC-MS, and LC-MS, and is supplied in small, pre-weighed pack sizes suitable for preparing standard solutions.
Catalog No. 50-002-81487 Supplier Medchemexpress LLC Supplier No. HYW393970S310MG
2′-Deoxyguanosine 5′-monophosphate-13C10,15N5 dilithium is an isotopically labeled nucleotide provided as the dilithium salt. It is intended for use as a tracer and internal standard in quantitative analyses by NMR, GC-MS, and LC-MS, and is supplied in small, pre-weighed pack sizes suitable for preparing standard solutions.