Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2′-deoxyguanosine 5′-monophosphate-d12 dilithium | 98.8% | 371.16 g/mol | C10D12Li2N5O7P | 1 MG
SDP

Catalog No. 5000281044
Encompass_Preferred

2′-Deoxyguanosine 5′-monophosphate-d12 dilithium is a deuterium-labeled deoxyribonucleotide provided as the dilithium salt for research use. It serves as an isotopically labeled standard for spectroscopic analyses and tracing experiments, offering reliable, high-purity material for analytical workflows.

  • Deuterium-labeled dGMP (d12) suitable for NMR and mass spectrometry.
  • Supplied as the dilithium salt for improved solubility and handling.
  • High purity (98.8%) for analytical confidence.
  • Molecular formula C10D12Li2N5O7P; molecular weight 371.16 g/mol.
  • Available in 1 mg vials; recommended formulation: 100 mM in 26.94 μL Tris HCl/D2O buffer pH 7.5.
  • Datasheet, certificate of analysis, and safety data sheet available.

Catalog No. 50-002-81044 Supplier Medchemexpress LLC Supplier No. HYW393970S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2′-Deoxyguanosine 5′-monophosphate-d12 dilithium is a deuterium-labeled deoxyribonucleotide provided as the dilithium salt for research use. It serves as an isotopically labeled standard for spectroscopic analyses and tracing experiments, offering reliable, high-purity material for analytical workflows.

  • Deuterium-labeled dGMP (d12) suitable for NMR and mass spectrometry.
  • Supplied as the dilithium salt for improved solubility and handling.
  • High purity (98.8%) for analytical confidence.
  • Molecular formula C10D12Li2N5O7P; molecular weight 371.16 g/mol.
  • Available in 1 mg vials; recommended formulation: 100 mM in 26.94 μL Tris HCl/D2O buffer pH 7.5.
  • Datasheet, certificate of analysis, and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.