Learn More
Medchemexpress LLC 2'-Deoxyguanosine 5'-monophosphate (disodium) | 33430-61-4 | 99.9% | 391.18 | 100 MG
SDP
Catalog No. 5000454674
Encompass_Preferred
2'-Deoxyguanosine 5'-monophosphate disodium (5′-dGMP) is a mononucleotide that has guanine as its nucleobase. This compound acts as a reactant in the analysis of self-assembling solutions and the nucleation/growth of G-quadruplexes. It is also involved in nucleophilic trapping and reductive alkylation processes. As a nucleic acid guanosine triphosphate (GTP) derivative, it serves as a nucleotide precursor in DNA synthesis.
  • Reactant in analysis of self-assembling solutions
  • Reactant in nucleation and growth of G-quadruplexes
  • Used in nucleophilic trapping
  • Used in reductive alkylation
  • Functions as an oxidizable target
  • Serves as a nucleotide precursor in DNA synthesis
  • Suitable for evaluating biologically relevant photosensitizing properties
Catalog No. 50-004-54674 Supplier Medchemexpress LLC Supplier No. HYW013159100MG
2'-Deoxyguanosine 5'-monophosphate disodium (5′-dGMP) is a mononucleotide that has guanine as its nucleobase. This compound acts as a reactant in the analysis of self-assembling solutions and the nucleation/growth of G-quadruplexes. It is also involved in nucleophilic trapping and reductive alkylation processes. As a nucleic acid guanosine triphosphate (GTP) derivative, it serves as a nucleotide precursor in DNA synthesis.
  • Reactant in analysis of self-assembling solutions
  • Reactant in nucleation and growth of G-quadruplexes
  • Used in nucleophilic trapping
  • Used in reductive alkylation
  • Functions as an oxidizable target
  • Serves as a nucleotide precursor in DNA synthesis
  • Suitable for evaluating biologically relevant photosensitizing properties