Learn More
Medchemexpress LLC 2'-Deoxyguanosine 5'-monophosphate disodium hydrate | 146877-98-7 | 99.0% | C10H14N5O7.H2O.2Na | 100 MG
SDP
Catalog No. 5000354139
Encompass_Preferred
2'-Deoxyguanosine 5'-monophosphate disodium hydrate (dGMP) is an oxidizable target of the photosensitizer pterin. It can be used to evaluate the photosensitizing properties of biopterins. Pterin causes a photosensitive reaction of dGMP under UV-A radiation, leading to DNA damage. This process involves two main mechanisms in vitro for the photosensitive oxidation of purine nucleotides by pterin.
  • A hydrogen abstraction reaction of electron transfer from dGMP to the triplet excited state of pterin (type I mechanism).
  • Interaction between dGMP and pterin produces singlet molecular oxygen (1O2) (type II mechanism).
Catalog No. 50-003-54139 Supplier Medchemexpress LLC Supplier No. HY145538100MG
2'-Deoxyguanosine 5'-monophosphate disodium hydrate (dGMP) is an oxidizable target of the photosensitizer pterin. It can be used to evaluate the photosensitizing properties of biopterins. Pterin causes a photosensitive reaction of dGMP under UV-A radiation, leading to DNA damage. This process involves two main mechanisms in vitro for the photosensitive oxidation of purine nucleotides by pterin.
  • A hydrogen abstraction reaction of electron transfer from dGMP to the triplet excited state of pterin (type I mechanism).
  • Interaction between dGMP and pterin produces singlet molecular oxygen (1O2) (type II mechanism).