Learn More
Medchemexpress LLC 2'-Deoxyguanosine 5'-monophosphate disodium hydrate | 146877-98-7 | 99.0% | C10H14N5O7.H2O.2Na | 50 MG
SDP
Catalog No. 5000354316
Encompass_Preferred
2'-Deoxyguanosine 5'-monophosphate (dGMP) disodium hydrate is an oxidizable target of pterin (PT) used to evaluate the photosensitizing properties of biopterins. Pterin induces a photosensitive reaction of dGMP under UV-A radiation, leading to DNA damage via two primary mechanisms: electron transfer (type I) and singlet molecular oxygen production (type II).
  • Oxidizable target of pterin (PT).
  • Evaluates photosensitizing properties of biopterins.
  • Induces DNA damage under UV-A radiation.
  • Functions through Type I (electron transfer) and Type II (singlet molecular oxygen production) mechanisms.
  • Relevant to cell cycle and DNA damage studies.
  • Involved in DNA/RNA synthesis research.
Catalog No. 50-003-54316 Supplier Medchemexpress LLC Supplier No. HY14553850MG
2'-Deoxyguanosine 5'-monophosphate (dGMP) disodium hydrate is an oxidizable target of pterin (PT) used to evaluate the photosensitizing properties of biopterins. Pterin induces a photosensitive reaction of dGMP under UV-A radiation, leading to DNA damage via two primary mechanisms: electron transfer (type I) and singlet molecular oxygen production (type II).
  • Oxidizable target of pterin (PT).
  • Evaluates photosensitizing properties of biopterins.
  • Induces DNA damage under UV-A radiation.
  • Functions through Type I (electron transfer) and Type II (singlet molecular oxygen production) mechanisms.
  • Relevant to cell cycle and DNA damage studies.
  • Involved in DNA/RNA synthesis research.