Learn More
Abcam 2'-Deo x yguanosine 5'-triphosphate [dGTP] trisodium salt, purine nucleoside triphosphate, 50MG
Catalog No. 502704583
Encompass_Preferred
MW 573.13 g/mol, Purity >98%. Purine nucleoside triphosphate used in many molecular biology applications such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques. Used in vivo for the synthesis of DNA via DNA polymerases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4583 Supplier Abcam Supplier No. AB14653650MG
MW 573.13 g/mol, Purity >98%. Purine nucleoside triphosphate used in many molecular biology applications such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques. Used in vivo for the synthesis of DNA via DNA polymerases.
The product is subject to the following: Abcam Restricted Use Statement