Learn More
Medchemexpress LLC 2'-Deoxyguanosine | 961-07-9 | 100.0% | 267.25 | 25 MG
SDP
Catalog No. 5000383946
Encompass_Preferred
2'-Deoxyguanosine is a purine nucleoside with various biological activities. It can induce DNA division in mouse thymus cells and acts as a potent cell division inhibitor in plant cells. It also demonstrates antiproliferative and antiviral effects against several cell lines and viruses.
  • Induces DNA division in mouse thymus cells.
  • Acts as a potent cell division inhibitor in plant cells.
  • Inhibits clonal growth of HL-60 and K562 leukemia cells.
  • Exhibits antiviral activity against varicella zoster virus (VZV) Oka strain.
  • Exhibits antiviral activity against human cytomegalovirus (HCMV) AD 169 strain.
  • Exhibits antiviral activity against herpes simplex virus-1 VR-3 strain.
  • Exhibits antiviral activity against herpes simplex virus-2 (HSV-2) Ms strain.
Catalog No. 50-003-83946 Supplier Medchemexpress LLC Supplier No. 25MGHY17563R
2'-Deoxyguanosine is a purine nucleoside with various biological activities. It can induce DNA division in mouse thymus cells and acts as a potent cell division inhibitor in plant cells. It also demonstrates antiproliferative and antiviral effects against several cell lines and viruses.
  • Induces DNA division in mouse thymus cells.
  • Acts as a potent cell division inhibitor in plant cells.
  • Inhibits clonal growth of HL-60 and K562 leukemia cells.
  • Exhibits antiviral activity against varicella zoster virus (VZV) Oka strain.
  • Exhibits antiviral activity against human cytomegalovirus (HCMV) AD 169 strain.
  • Exhibits antiviral activity against herpes simplex virus-1 VR-3 strain.
  • Exhibits antiviral activity against herpes simplex virus-2 (HSV-2) Ms strain.