Learn More
Medchemexpress LLC 2'-Deoxyguanosine | 961-07-9 | 267.25 | 1 G
SDP
Catalog No. 5000386439
Encompass_Preferred
2'-Deoxyguanosine is a purine nucleoside with a variety of biological activities. It can induce DNA division in mouse thymus cells and is a potent cell division inhibitor in plant cells. In vitro, it inhibits the clonal growth of HL-60 and K562 leukemia cells with IC50 values of 50 and 80 μM, respectively.
  • Classified as an endogenous metabolite related to cancer
  • Analytical standard intended for research and analytical applications (for standard version HY-17563R)
Catalog No. 50-003-86439 Supplier Medchemexpress LLC Supplier No. HY175631G
2'-Deoxyguanosine is a purine nucleoside with a variety of biological activities. It can induce DNA division in mouse thymus cells and is a potent cell division inhibitor in plant cells. In vitro, it inhibits the clonal growth of HL-60 and K562 leukemia cells with IC50 values of 50 and 80 μM, respectively.
  • Classified as an endogenous metabolite related to cancer
  • Analytical standard intended for research and analytical applications (for standard version HY-17563R)