Learn More
Medchemexpress LLC 2'-Deoxyguanosine | 961-07-9 | 99.21% | 267.24 | 100 MG
SDP
Catalog No. 5000387158
Encompass_Preferred
2'-Deoxyguanosine (Standard) is an analytical standard of 2'-Deoxyguanosine, intended for research and analytical applications. It is a purine nucleoside with various biological activities, including inducing DNA division in mouse thymus cells and acting as a potent cell division inhibitor in plant cells. It also inhibits the clonal growth of HL-60 and K562 leukemia cells with IC50 values of 50 and 80 μM, respectively.
  • Analytical standard for 2'-Deoxyguanosine.
  • Intended for research and analytical applications.
  • Purine nucleoside with various biological activities.
  • Induces DNA division in mouse thymus cells.
  • Potent cell division inhibitor in plant cells.
  • Inhibits clonal growth of HL-60 and K562 leukemia cells (IC50 values of 50 and 80 μM).
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-87158 Supplier Medchemexpress LLC Supplier No. HY17563R100MG
2'-Deoxyguanosine (Standard) is an analytical standard of 2'-Deoxyguanosine, intended for research and analytical applications. It is a purine nucleoside with various biological activities, including inducing DNA division in mouse thymus cells and acting as a potent cell division inhibitor in plant cells. It also inhibits the clonal growth of HL-60 and K562 leukemia cells with IC50 values of 50 and 80 μM, respectively.
  • Analytical standard for 2'-Deoxyguanosine.
  • Intended for research and analytical applications.
  • Purine nucleoside with various biological activities.
  • Induces DNA division in mouse thymus cells.
  • Potent cell division inhibitor in plant cells.
  • Inhibits clonal growth of HL-60 and K562 leukemia cells (IC50 values of 50 and 80 μM).
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.