Learn More
Medchemexpress LLC 2'-Deoxyguanosine | 961-07-9 | 99.9% | 267.25 | 10 G
SDP
Catalog No. 5000386373
Encompass_Preferred
2'-Deoxyguanosine is a purine nucleoside with diverse biological activities, known to induce DNA division in mouse thymus cells and act as a potent cell division inhibitor in plant cells. It also demonstrates inhibitory effects on the clonal growth of HL-60 and K562 leukemia cells, with observed IC50 values of 50 and 80 μM, respectively. This compound typically appears as a solid, ranging from white to off-white in color.
  • Endogenous metabolite
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
Catalog No. 50-003-86373 Supplier Medchemexpress LLC Supplier No. HY1756310G
2'-Deoxyguanosine is a purine nucleoside with diverse biological activities, known to induce DNA division in mouse thymus cells and act as a potent cell division inhibitor in plant cells. It also demonstrates inhibitory effects on the clonal growth of HL-60 and K562 leukemia cells, with observed IC50 values of 50 and 80 μM, respectively. This compound typically appears as a solid, ranging from white to off-white in color.
  • Endogenous metabolite
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation