Learn More
Medchemexpress LLC 2'-Deoxyguanosine | 961-07-9 | >99.9% | 267.25 | 5 G
SDP
Catalog No. 5000386301
Encompass_Preferred
2'-Deoxyguanosine is a purine nucleoside with diverse biological activities. It can induce DNA division in mouse thymus cells and acts as a potent cell division inhibitor in plant cells. It inhibits the clonal growth of HL-60 and K562 leukemia cells with IC50 values of 50 and 80 μM, respectively. This product is intended for research use only.
  • Induces DNA division in mouse thymus cells.
  • Acts as a potent cell division inhibitor in plant cells.
  • Inhibits clonal growth of HL-60 and K562 leukemia cells.
  • Intended for research applications.
Catalog No. 50-003-86301 Supplier Medchemexpress LLC Supplier No. HY175635G
2'-Deoxyguanosine is a purine nucleoside with diverse biological activities. It can induce DNA division in mouse thymus cells and acts as a potent cell division inhibitor in plant cells. It inhibits the clonal growth of HL-60 and K562 leukemia cells with IC50 values of 50 and 80 μM, respectively. This product is intended for research use only.
  • Induces DNA division in mouse thymus cells.
  • Acts as a potent cell division inhibitor in plant cells.
  • Inhibits clonal growth of HL-60 and K562 leukemia cells.
  • Intended for research applications.