Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyguanosine | 961-07-9 | MFCD00080300 | 99.2% | 267.24 g/mol | C10H13N5O4 | 10 MG
SDP

Catalog No. 5000298754
Encompass_Preferred

2'-Deoxyguanosine (standard) is the analytical standard of 2'-deoxyguanosine, a purine nucleoside used as an analytical reference and research reagent. Supplied as a high-purity solid, it is intended for analytical assays, method development, and quantitative reference applications.

  • Analytical standard suitable for quantitative assays and calibration.
  • High purity (99.2%) for reliable analytical results.
  • Molecular weight 267.24 g/mol; formula C10H13N5O4 for accurate identification.
  • Available in small pack sizes for method development and trace analyses.
  • Supplied with certificate of analysis and recommended storage guidance.

Catalog No. 50-002-98754 Supplier Medchemexpress LLC Supplier No. HY17563R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyguanosine (standard) is the analytical standard of 2'-deoxyguanosine, a purine nucleoside used as an analytical reference and research reagent. Supplied as a high-purity solid, it is intended for analytical assays, method development, and quantitative reference applications.

  • Analytical standard suitable for quantitative assays and calibration.
  • High purity (99.2%) for reliable analytical results.
  • Molecular weight 267.24 g/mol; formula C10H13N5O4 for accurate identification.
  • Available in small pack sizes for method development and trace analyses.
  • Supplied with certificate of analysis and recommended storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.