Learn More
Bioworld 2'-Deoxyguanosine Monohydrate, 250 mg
Supplier:  Bioworld 404900121
Encompass_Preferred
Nucleoside analogSpecifications:Ultra Pure GradeEm (252nm, phosphate buffer, pH 7.0): >13,300 Appearance: Powder.ChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: >300 °C (dec.)(lit.)
Catalog No. 50-255-898