Learn More
Abcam 2'-Deo x yguanosine monohydrate, purine nucleoside, 1G
Catalog No. 502704594
Encompass_Preferred
MW 285.2 g/mol, Purity >98%. Purine nucleoside that upon sequential phosphoylation forms dGTP which is used by DNA polymerases and reverse transcriptases to synthesise DNA.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4594 Supplier Abcam Supplier No. AB1465261G
MW 285.2 g/mol, Purity >98%. Purine nucleoside that upon sequential phosphoylation forms dGTP which is used by DNA polymerases and reverse transcriptases to synthesise DNA.
The product is subject to the following: Abcam Restricted Use Statement