Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxyguanosine monohydrate | 312693-72-4 | 99.7% | 10 G
SDP

Catalog No. 5000410266
Encompass_Preferred

2'-Deoxyguanosine monohydrate is a nucleoside composed of guanine and 2'-deoxyribose, and it is one of the basic structural units of DNA. The oxidation products of 2'-Deoxyguanosine monohydrate may participate in DNA damage, which in turn may affect gene expression or induce cancer.

  • Composed of guanine and 2'-deoxyribose.
  • Basic structural unit of DNA.
  • Oxidation products may participate in DNA damage.
  • Can affect gene expression or induce cancer.
  • High purity of 99.70%.
  • Ships at room temperature.
  • Store solid at 4°C, sealed, away from moisture.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture.

Catalog No. 50-004-10266 Supplier Medchemexpress LLC Supplier No. HYD018510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyguanosine monohydrate is a nucleoside composed of guanine and 2'-deoxyribose, and it is one of the basic structural units of DNA. The oxidation products of 2'-Deoxyguanosine monohydrate may participate in DNA damage, which in turn may affect gene expression or induce cancer.

  • Composed of guanine and 2'-deoxyribose.
  • Basic structural unit of DNA.
  • Oxidation products may participate in DNA damage.
  • Can affect gene expression or induce cancer.
  • High purity of 99.70%.
  • Ships at room temperature.
  • Store solid at 4°C, sealed, away from moisture.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.