Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxyguanosine monohydrate | 312693-72-4 | 99.7% | 10 UMOL
SDP

Catalog No. 5000410944
Encompass_Preferred

2'-Deoxyguanosine monohydrate is a nucleoside composed of guanine and 2'-deoxyribose, serving as a fundamental structural unit of DNA. Its oxidation products may contribute to DNA damage, which can consequently affect gene expression or induce cancer. This product is intended for research use only.

  • Molecular weight: 285.26
  • Formula: C10H15N5O5
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-004-10944 Supplier Medchemexpress LLC Supplier No. HYD018510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyguanosine monohydrate is a nucleoside composed of guanine and 2'-deoxyribose, serving as a fundamental structural unit of DNA. Its oxidation products may contribute to DNA damage, which can consequently affect gene expression or induce cancer. This product is intended for research use only.

  • Molecular weight: 285.26
  • Formula: C10H15N5O5
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.