Learn More
Medchemexpress LLC 2'-Deoxyguanosine monohydrate | 312693-72-4 | 99.7% | 5 G
SDP
Catalog No. 5000409830
Encompass_Preferred
2'-Deoxyguanosine monohydrate is a nucleoside composed of guanine and 2'-deoxyribose, and it is one of the basic structural units of DNA.
  • The oxidation products may participate in DNA damage.
  • May affect gene expression or induce cancer.
Catalog No. 50-004-09830 Supplier Medchemexpress LLC Supplier No. HYD01855G
2'-Deoxyguanosine monohydrate is a nucleoside composed of guanine and 2'-deoxyribose, and it is one of the basic structural units of DNA.
  • The oxidation products may participate in DNA damage.
  • May affect gene expression or induce cancer.