Learn More
Medchemexpress LLC 2'-Deoxyguanosine (standard) | 961-07-9 | MFCD00080300 | 99.2% | 267.24 | 50 MG
SDP
Catalog No. 5000387159
Encompass_Preferred
2'-Deoxyguanosine (Standard) is the analytical standard for 2'-Deoxyguanosine. It is intended for research and analytical applications. This purine nucleoside exhibits diverse biological activities, including inducing DNA division in mouse thymus cells and acting as a potent cell division inhibitor in plant cells. In vitro, it inhibits the clonal growth of HL-60 and K562 leukemia cells with IC50 values of 50 and 80 μM, respectively.
  • Analytical standard for 2'-deoxyguanosine
  • Intended for research and analytical applications
  • Induces DNA division in mouse thymus cells
  • Potent cell division inhibitor in plant cells
  • Inhibits clonal growth of HL-60 and K562 leukemia cells
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-87159 Supplier Medchemexpress LLC Supplier No. HY17563R50MG
2'-Deoxyguanosine (Standard) is the analytical standard for 2'-Deoxyguanosine. It is intended for research and analytical applications. This purine nucleoside exhibits diverse biological activities, including inducing DNA division in mouse thymus cells and acting as a potent cell division inhibitor in plant cells. In vitro, it inhibits the clonal growth of HL-60 and K562 leukemia cells with IC50 values of 50 and 80 μM, respectively.
  • Analytical standard for 2'-deoxyguanosine
  • Intended for research and analytical applications
  • Induces DNA division in mouse thymus cells
  • Potent cell division inhibitor in plant cells
  • Inhibits clonal growth of HL-60 and K562 leukemia cells
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications