Learn More
Cayman Chemical 2'-Deoxyguanosine-5'-O-triphosphate (sodium salt), Purity: ≥95%, CAS: 93919-41-6, 10 mg
SDP
Catalog No. 502911403
Encompass_Preferred
A nucleoside triphosphate; has commonly been used in PCR reaction mixtures as a substrate for DNA synthesis by DNA polymerases; can be oxidized to 8-hydroxy-dGTP and 8-oxo-7,8-GTP, which can be misincorporated into DNA
Catalog No. 50-291-1403 Supplier Cayman Chemical Supplier No. 3577110 MG
A nucleoside triphosphate; has commonly been used in PCR reaction mixtures as a substrate for DNA synthesis by DNA polymerases; can be oxidized to 8-hydroxy-dGTP and 8-oxo-7,8-GTP, which can be misincorporated into DNA