Learn More
Cayman Chemical 2-DeoxygunosIn-5-O-trIpho 50mg
SDP
Catalog No. 502910261
Encompass_Preferred
A nucleoside triphosphate; has commonly been used in PCR reaction mixtures as a substrate for DNA synthesis by DNA polymerases; can be oxidized to 8-hydroxy-dGTP and 8-oxo-7,8-GTP, which can be misincorporated into DNA
Catalog No. 50-291-0261 Supplier Cayman Chemical Supplier No. 3577150 MG
A nucleoside triphosphate; has commonly been used in PCR reaction mixtures as a substrate for DNA synthesis by DNA polymerases; can be oxidized to 8-hydroxy-dGTP and 8-oxo-7,8-GTP, which can be misincorporated into DNA