Learn More
Bioworld 2'-Deoxythymidine, 25 g
Supplier:  Bioworld 404900162
Encompass_Preferred
It is a pyrimidine deoxynucleoside.It is the DNA nucleoside T, which pairs with deoxyadenosine (A) in double-stranded DNA.It can be phosphorylated with one, two or three phosphoric acid groups, creating respectively deoxythymidine mono,di or triphosphate.It is non-toxic and as part of one of the four nucleotides in DNA it is a naturally occurring compound that exists in all living organisms and DNA viruses. Specifications:Ultra Pure GradeEm (267nm, phosphate buffer, pH 7.0): >9,200Appearance: White Crystalline PowderChEBI : Not available.Crystal_Structure : Not available.Density : 1.452 g/cm3InChI : Not available.InChIKey : Not available.IUPAC_Name: 1-[4-Hydroxy-5-(Hydroxymethyl)Oxolan-2-Yl]-5-Methyl-Pyrimidine-2,4-DioneMelting Point: 186-188 °C(lit.)RTECS Number: XP2071000
Catalog No. 50-255-900