Learn More
Abcam 2'-Deo x ythymidine 5'-triphosphate [dTTP] trisodium salt, nucleoside triphosphate, 50MG
Catalog No. 502704579
Encompass_Preferred
MW 548.12 g/mol, Purity >98%. Nucleoside triphosphate used in many molecular biology applications such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4579 Supplier Abcam Supplier No. AB14654050MG
MW 548.12 g/mol, Purity >98%. Nucleoside triphosphate used in many molecular biology applications such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques.
The product is subject to the following: Abcam Restricted Use Statement