Learn More
Cayman Chemical 2'-Deoxyuridine, Purity: ≥98%, CAS: 951-78-0, 1 g
SDP
Catalog No. 502939266
Encompass_Preferred
A pyrimidine nucleoside; has been used as a precursor in the synthesis of antiviral deoxyuridine derivatives, including idoxuridine
Catalog No. 50-293-9266 Supplier Cayman Chemical Supplier No. 278031 G
A pyrimidine nucleoside; has been used as a precursor in the synthesis of antiviral deoxyuridine derivatives, including idoxuridine