Learn More
Cayman Chemical 2'-Deoxyuridine, Purity: ≥98%, CAS: 951-78-0, 500 mg
SDP
Catalog No. 502938701
Encompass_Preferred
A pyrimidine nucleoside; has been used as a precursor in the synthesis of antiviral deoxyuridine derivatives, including idoxuridine
Catalog No. 50-293-8701 Supplier Cayman Chemical Supplier No. 27803500 MG
A pyrimidine nucleoside; has been used as a precursor in the synthesis of antiviral deoxyuridine derivatives, including idoxuridine