Learn More
Cayman Chemical 2′-Deoxyuridine-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 40632-23-3, 5 mg
SDP
Catalog No. 502911885
Encompass_Preferred
An internal standard for the quantification of 2’-deoxyuridine by GC- or LC-MS
Catalog No. 50-291-1885 Supplier Cayman Chemical Supplier No. 363235 MG
An internal standard for the quantification of 2’-deoxyuridine by GC- or LC-MS