Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxyuridine-1'-13C | 478510-85-9 | 1MG
SDP

Catalog No. 5000410677
Encompass_Preferred

2'-Deoxyuridine-1'-13C is a stable isotope-labeled compound with the molecular formula C813CH12N2O5 and a molecular weight of 229.19. It appears as a white to off-white solid with an isotopic enrichment of 99%. This product is intended for research use only and has not been fully validated for medical applications.

  • White to off-white solid appearance
  • Isotopic enrichment of 99%
  • Molecular formula C813CH12N2O5
  • Intended for research use only
  • Store powder at -20°C for up to 3 years or at 4°C for up to 2 years
  • Store in solvent at -80°C for 6 months or at -20°C for 1 month

Catalog No. 50-004-10677 Supplier Medchemexpress LLC Supplier No. HYD0186S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyuridine-1'-13C is a stable isotope-labeled compound with the molecular formula C813CH12N2O5 and a molecular weight of 229.19. It appears as a white to off-white solid with an isotopic enrichment of 99%. This product is intended for research use only and has not been fully validated for medical applications.

  • White to off-white solid appearance
  • Isotopic enrichment of 99%
  • Molecular formula C813CH12N2O5
  • Intended for research use only
  • Store powder at -20°C for up to 3 years or at 4°C for up to 2 years
  • Store in solvent at -80°C for 6 months or at -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.