Learn More
Medchemexpress LLC HY-D0186 100mg Medchemexpress, 2'-Deoxyuridine CAS:951-78-0 Purity:>98%
SDP
Catalog No. 501933154
Encompass_Preferred
Medchemexpress, HY-D0186 100mg 2'-Deoxyuridine CAS:951-78-0 2'-Deoxyuridine could increase chromosome breakage and results in a decreased thymidylate synthetase activity. A known use of 2'-Deoxyuridine is as a precursor in the synthesis of Edoxudine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3154 Supplier Medchemexpress LLC Supplier No. HYD0186100MG
Medchemexpress, HY-D0186 100mg 2'-Deoxyuridine CAS:951-78-0 2'-Deoxyuridine could increase chromosome breakage and results in a decreased thymidylate synthetase activity. A known use of 2'-Deoxyuridine is as a precursor in the synthesis of Edoxudine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.