Learn More
Medchemexpress LLC 2'-Deoxyuridine-13C,15N2 | 369656-76-8 | 98.1% | 1 MG
SDP
Catalog No. 5000409831
Encompass_Preferred
2'-Deoxyuridine-13C,15N2 is the 13C and 15N labeled 2'-Deoxyuridine, a compound that can increase chromosome breakage and decrease thymidylate synthetase activity. It is also known as a precursor in the synthesis of Edoxudine.
  • Can increase chromosome breakage.
  • Results in decreased thymidylate synthetase activity.
  • Precursor in the synthesis of Edoxudine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09831 Supplier Medchemexpress LLC Supplier No. HYD0186S71MG
2'-Deoxyuridine-13C,15N2 is the 13C and 15N labeled 2'-Deoxyuridine, a compound that can increase chromosome breakage and decrease thymidylate synthetase activity. It is also known as a precursor in the synthesis of Edoxudine.
  • Can increase chromosome breakage.
  • Results in decreased thymidylate synthetase activity.
  • Precursor in the synthesis of Edoxudine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.