Learn More
Selleck Chemical LLC 2-deoxyuridine
Supplier: Selleck Chemical LLC S550525MG
2 -deoxyuridine (dU 2 -dU) is frequently halogenated to create thymidine analogues useful for studies of DNA synthesis and degradation mechanisms