Learn More
Selleck Chemical LLC 2-deoxyuridine
SDP
Catalog No. 5000003823
Encompass_Preferred
2 -deoxyuridine (dU 2 -dU) is frequently halogenated to create thymidine analogues useful for studies of DNA synthesis and degradation mechanisms
Catalog No. 50-000-03823 Supplier Selleck Chemical LLC Supplier No. S55051G
2 -deoxyuridine (dU 2 -dU) is frequently halogenated to create thymidine analogues useful for studies of DNA synthesis and degradation mechanisms