Learn More
Apexbio Technology LLC 2-Deoxyuridine 5-monophosphate disodium 42155-08-8 250mg
SDP
Catalog No. 5000089294
Encompass_Preferred
2-Deoxyuridine 5-monophosphate disodium (CAS 42155-08-8) is a nucleotide analog recognized as a natural substrate for thymidylate synthase (TS) TS catalyzes the methylation of this compound utilizing 5 10-methylenetetrahydrofolate as a methyl donor to generate deoxythymidine 5-monophosphate (dTMP) This enzymatic conversion constitutes an essential step in the de novo synthesis of thymidine nucleotides required for DNA replication 2-Deoxyuridine 5-monophosphate disodium is widely employed in studies exploring DNA synthesis the thymidylate biosynthetic pathway and the regulation of nucleotide metabolism
Catalog No. 50-000-89294 Supplier Apexbio Technology LLC Supplier No. B8925250
2-Deoxyuridine 5-monophosphate disodium (CAS 42155-08-8) is a nucleotide analog recognized as a natural substrate for thymidylate synthase (TS) TS catalyzes the methylation of this compound utilizing 5 10-methylenetetrahydrofolate as a methyl donor to generate deoxythymidine 5-monophosphate (dTMP) This enzymatic conversion constitutes an essential step in the de novo synthesis of thymidine nucleotides required for DNA replication 2-Deoxyuridine 5-monophosphate disodium is widely employed in studies exploring DNA synthesis the thymidylate biosynthetic pathway and the regulation of nucleotide metabolism