Learn More
Medchemexpress LLC 2'-Deoxyuridine 5'-monophosphate disodium | 42155-08-8 | CB9482517 | 100.0% | 352.15 | 250 MG
SDP
Catalog No. 5000453271
Encompass_Preferred
2'-Deoxyuridine 5'-monophosphate disodium (dUMP disodium) is a deoxynucleotide and an endogenous metabolite. It is reductively methylated to dTMP, a nucleotide essential for DNA synthesis. This compound exhibits anti-bacterial activity against *Helicobacter pylori* by inhibiting HpThyX enzyme activities. It is provided as a solid, white to off-white in color, and is intended for research use only.
  • Deoxynucleotide
  • Reductively methylated to dTMP
  • Required for DNA synthesis
  • Exhibits anti-bacterial activity
  • Solid appearance
  • White to off-white color
  • Endogenous metabolite
  • For research use only
Catalog No. 50-004-53271 Supplier Medchemexpress LLC Supplier No. 250MGHYW011142
2'-Deoxyuridine 5'-monophosphate disodium (dUMP disodium) is a deoxynucleotide and an endogenous metabolite. It is reductively methylated to dTMP, a nucleotide essential for DNA synthesis. This compound exhibits anti-bacterial activity against *Helicobacter pylori* by inhibiting HpThyX enzyme activities. It is provided as a solid, white to off-white in color, and is intended for research use only.
  • Deoxynucleotide
  • Reductively methylated to dTMP
  • Required for DNA synthesis
  • Exhibits anti-bacterial activity
  • Solid appearance
  • White to off-white color
  • Endogenous metabolite
  • For research use only