Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyuridine-5'-triphosphate trisodium | 102814-08-4 | MFCD00084701 | 99.7% | 534.09 g·mol⁻1 | C9H12N2Na3O14P3 | 1 ML
SDP

Catalog No. 5000300168
Encompass_Preferred

dUTP (trisodium) is the trisodium salt of 2'-deoxyuridine-5'-triphosphate supplied as a 100 mM PCR-grade aqueous solution. It is used as a nucleotide substrate for DNA synthesis, PCR-based assays, and labeling reactions, and is provided with supporting documentation for quality control.

  • PCR-grade 100 mM aqueous solution suitable for enzymatic reactions.
  • High purity (HPLC 99.74%) for reliable incorporation by polymerases.
  • Colorless to light yellow liquid for easy visual inspection.
  • Molecular weight 534.09 g·mol⁻1 and trisodium salt formulation.
  • Supplied with COA and SDS for traceability and compliance.
  • Recommended storage: in solvent -80°C (up to 6 months) or -20°C (short term).

Catalog No. 50-003-00168 Supplier Medchemexpress LLC Supplier No. HYW017068B1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

dUTP (trisodium) is the trisodium salt of 2'-deoxyuridine-5'-triphosphate supplied as a 100 mM PCR-grade aqueous solution. It is used as a nucleotide substrate for DNA synthesis, PCR-based assays, and labeling reactions, and is provided with supporting documentation for quality control.

  • PCR-grade 100 mM aqueous solution suitable for enzymatic reactions.
  • High purity (HPLC 99.74%) for reliable incorporation by polymerases.
  • Colorless to light yellow liquid for easy visual inspection.
  • Molecular weight 534.09 g·mol⁻1 and trisodium salt formulation.
  • Supplied with COA and SDS for traceability and compliance.
  • Recommended storage: in solvent -80°C (up to 6 months) or -20°C (short term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.