Learn More
Medchemexpress LLC 2'-deoxyuridine-5'-triphosphate trisodium salt | 102814-08-4 | MFCD00084701 | 99.7% | 534.09 | C9H12N2Na3O14P3 | 5 MG
SDP
Catalog No. 5000300162
Encompass_Preferred
dUTP trisodium (2'-deoxyuridine-5'-triphosphate trisodium salt) is the trisodium salt of dUTP, a deoxynucleotide triphosphate used as a dTTP substitute in PCR and related enzymatic applications to produce uracil-containing DNA. Supplied as a solid with high reported purity, it is intended for workflows that require uracil incorporation or UDG-based carryover prevention.
  • High purity (99.74%) suitable for sensitive enzymatic reactions.
  • Provided as a white to off-white solid, 5 mg pack size.
  • Trisodium salt form for stable handling and solubility.
  • Compatible with PCR and RT-PCR to produce uracil-containing PCR products.
  • Supports uracil-DNA glycosylase (UDG)-based carryover prevention.
  • Store sealed, away from moisture; in solution: -80°C (up to 6 months), -20°C (up to 1 month).
Catalog No. 50-003-00162 Supplier Medchemexpress LLC Supplier No. HYW0170685MG
dUTP trisodium (2'-deoxyuridine-5'-triphosphate trisodium salt) is the trisodium salt of dUTP, a deoxynucleotide triphosphate used as a dTTP substitute in PCR and related enzymatic applications to produce uracil-containing DNA. Supplied as a solid with high reported purity, it is intended for workflows that require uracil incorporation or UDG-based carryover prevention.
  • High purity (99.74%) suitable for sensitive enzymatic reactions.
  • Provided as a white to off-white solid, 5 mg pack size.
  • Trisodium salt form for stable handling and solubility.
  • Compatible with PCR and RT-PCR to produce uracil-containing PCR products.
  • Supports uracil-DNA glycosylase (UDG)-based carryover prevention.
  • Store sealed, away from moisture; in solution: -80°C (up to 6 months), -20°C (up to 1 month).