Learn More
Medchemexpress LLC 2'-Deoxyuridine | 951-78-0 | 100.0% | 10 MM 1 ML
SDP
Catalog No. 5000409924
Encompass_Preferred
2'-Deoxyuridine is a brain-penetrant pyrimidines nucleotide associated with nervous system diseases. It can increase chromosome breakage and decrease thymidylate synthetase activity. It serves as a precursor in the synthesis of Edoxudine and is an analogue of 5-ethynyl-2'-deoxyuridine. Additionally, 2'-deoxyuridine reduces microglial activation and improves oxidative stress damage by modulating glycolytic metabolism on Aβ25-35-induced brain injury, showing promise for Alzheimer's disease (AD) research.
  • Brain-penetrant pyrimidines nucleotide associated with nervous system diseases.
  • Increases chromosome breakage.
  • Decreases thymidylate synthetase activity.
  • Reduces microglial activation.
  • Improves oxidative stress damage by modulating glycolytic metabolism on Aβ25-35-induced brain injury.
  • Promising for Alzheimer's disease (AD) research.
Catalog No. 50-004-09924 Supplier Medchemexpress LLC Supplier No. HYD018610MM/1ML
2'-Deoxyuridine is a brain-penetrant pyrimidines nucleotide associated with nervous system diseases. It can increase chromosome breakage and decrease thymidylate synthetase activity. It serves as a precursor in the synthesis of Edoxudine and is an analogue of 5-ethynyl-2'-deoxyuridine. Additionally, 2'-deoxyuridine reduces microglial activation and improves oxidative stress damage by modulating glycolytic metabolism on Aβ25-35-induced brain injury, showing promise for Alzheimer's disease (AD) research.
  • Brain-penetrant pyrimidines nucleotide associated with nervous system diseases.
  • Increases chromosome breakage.
  • Decreases thymidylate synthetase activity.
  • Reduces microglial activation.
  • Improves oxidative stress damage by modulating glycolytic metabolism on Aβ25-35-induced brain injury.
  • Promising for Alzheimer's disease (AD) research.