Learn More
Apexbio Technology LLC 2-Deoxyuridine 951-78-0 500mg
SDP
Catalog No. 5000091697
Encompass_Preferred
2-Deoxyuridine (CAS 951-78-0) is a pyrimidine nucleoside analog in which the 2 -hydroxyl group of uridine is absent In biological systems it can be incorporated into DNA in place of thymidine thereby influencing DNA structure and metabolism The compound serves as a precursor for halogenated nucleoside analogues widely utilized in studies examining DNA replication and degradation pathways 2-Deoxyuridine is frequently employed in molecular biology research to investigate DNA synthesis mechanisms and to facilitate the development of labeled nucleic acid probes
Catalog No. 50-000-91697 Supplier Apexbio Technology LLC Supplier No. B7866500
2-Deoxyuridine (CAS 951-78-0) is a pyrimidine nucleoside analog in which the 2 -hydroxyl group of uridine is absent In biological systems it can be incorporated into DNA in place of thymidine thereby influencing DNA structure and metabolism The compound serves as a precursor for halogenated nucleoside analogues widely utilized in studies examining DNA replication and degradation pathways 2-Deoxyuridine is frequently employed in molecular biology research to investigate DNA synthesis mechanisms and to facilitate the development of labeled nucleic acid probes