Learn More
Medchemexpress LLC 2'-deoxyuridine | 951-78-0 | MFCD00006527 | 228.20 g/mol | C9H12N2O5 | 10 MG
SDP
Catalog No. 5000296720
Encompass_Preferred
2'-Deoxyuridine is a pyrimidine 2'-deoxyribonucleoside used in nucleic acid and metabolic research. It is applied as an analytical standard and reagent for studies of DNA synthesis, repair, and related enzymatic assays. Supplied as a 10 mg research sample.
  • Endogenous metabolite used in biochemical and metabolic studies.
  • Suitable as an analytical standard for chromatographic methods.
  • Applicable in nucleic acid synthesis and enzymatic assays.
  • Solid powder form simplifies weighing and formulation.
  • Store and handle according to the safety data sheet and laboratory practices.
Catalog No. 50-002-96720 Supplier Medchemexpress LLC Supplier No. HYD018610MG
2'-Deoxyuridine is a pyrimidine 2'-deoxyribonucleoside used in nucleic acid and metabolic research. It is applied as an analytical standard and reagent for studies of DNA synthesis, repair, and related enzymatic assays. Supplied as a 10 mg research sample.
  • Endogenous metabolite used in biochemical and metabolic studies.
  • Suitable as an analytical standard for chromatographic methods.
  • Applicable in nucleic acid synthesis and enzymatic assays.
  • Solid powder form simplifies weighing and formulation.
  • Store and handle according to the safety data sheet and laboratory practices.