Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyuridine | 951-78-0 | MFCD00006527 | 99.2% | 228.20 g/mol | C9H12N2O5 | 10 MG
SDP

Catalog No. 5000297516
Encompass_Preferred

2'-Deoxyuridine standard supplied as an analytical reference for research and analytical applications. It serves as a reference compound for biochemical assays, method development, and studies of nucleotide metabolism and neurodegenerative disease models.

  • Suitable as an analytical reference standard for biochemical and pharmaceutical assays.
  • High purity, 99.2%.
  • Consistent chemical identity: C9H12N2O5, MW 228.20 g/mol.
  • Available in small quantities for analytical use, including 10 mg vials.
  • Shipped at room temperature in continental US; store per certificate of analysis.

Catalog No. 50-002-97516 Supplier Medchemexpress LLC Supplier No. HYD0186R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyuridine standard supplied as an analytical reference for research and analytical applications. It serves as a reference compound for biochemical assays, method development, and studies of nucleotide metabolism and neurodegenerative disease models.

  • Suitable as an analytical reference standard for biochemical and pharmaceutical assays.
  • High purity, 99.2%.
  • Consistent chemical identity: C9H12N2O5, MW 228.20 g/mol.
  • Available in small quantities for analytical use, including 10 mg vials.
  • Shipped at room temperature in continental US; store per certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.