Learn More
Medchemexpress LLC 2'-Deoxyuridine (Standard) | 951-78-0 | 99.2% | 25 MG
SDP
Catalog No. 5000413424
Encompass_Preferred
2'-Deoxyuridine (Standard) is the analytical standard of 2'-Deoxyuridine, intended for research and analytical applications. It is a brain-penetrant pyrimidines nucleotide associated with nervous system diseases. It can increase chromosome breakage and decrease thymidylate synthetase activity. It reduces microglial activation and improves oxidative stress damage by modulating glycolytic metabolism on Aβ25-35-induced brain injury, showing promise for Alzheimer's disease (AD) research.
  • Used as an analytical standard.
  • Precursor in the synthesis of Edoxudine.
  • Analogue of 5-ethynyl-2'-deoxyuridine.
  • Reduces microglial activation.
  • Improves oxidative stress damage.
  • Promising for Alzheimer's disease research.
  • Utilized in HPLC, GC, and MS research experiments.
Catalog No. 50-004-13424 Supplier Medchemexpress LLC Supplier No. 25MGHYD0186R
2'-Deoxyuridine (Standard) is the analytical standard of 2'-Deoxyuridine, intended for research and analytical applications. It is a brain-penetrant pyrimidines nucleotide associated with nervous system diseases. It can increase chromosome breakage and decrease thymidylate synthetase activity. It reduces microglial activation and improves oxidative stress damage by modulating glycolytic metabolism on Aβ25-35-induced brain injury, showing promise for Alzheimer's disease (AD) research.
  • Used as an analytical standard.
  • Precursor in the synthesis of Edoxudine.
  • Analogue of 5-ethynyl-2'-deoxyuridine.
  • Reduces microglial activation.
  • Improves oxidative stress damage.
  • Promising for Alzheimer's disease research.
  • Utilized in HPLC, GC, and MS research experiments.