Learn More
Medchemexpress LLC HY-135009 100mg Medchemexpress, 2-Di-1-ASP CAS:2156-29-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135009100MG
Encompass_Preferred
Medchemexpress, HY-135009 100mg 2-Di-1-ASP CAS:2156-29-8 2-Di-1-ASP (Compound 18a) is a mono-stryryl dye, and widely used as mitochondrial stain and groove-binding fluorescent probes for double-stranded DNA. 2-Di-1-ASP is selective for G-quadruplex (G4) and double-stranded DNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9857