Learn More
Strem, An Ascensus Company CAS# 564483-19-8. 500mg. 2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% t-BuXPhos. MFCD06411306
SDP
Catalog No. 5090110050
Encompass
CAS# 564483-19-8. 500mg. 2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% t-BuXPhos. MFCD06411306. Molecular Weight: 424.64. Molecular Formula: C29H45P. Color/form: white xtl. Strem# 15-1052. http://www.strem.com/catalog/v/15-1052/
Catalog No. 50-901-10050 Supplier Strem, An Ascensus Company Supplier No. 151052500MG
CAS# 564483-19-8. 500mg. 2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% t-BuXPhos. MFCD06411306. Molecular Weight: 424.64. Molecular Formula: C29H45P. Color/form: white xtl. Strem# 15-1052. http://www.strem.com/catalog/v/15-1052/